4-Bromo-2-fluoro-5-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

046604-1G
101876-242EA 281.75 USD
101876-242
4-Bromo-2-fluoro-5-nitrobenzaldehyde ≥95%
4-Bromo-2-fluoro-5-nitrobenzaldehyde

Hazard: Irritant

Formula: C₇H₃BrFNO₃
MW: 248.0060 g/mol
Melting Pt: 47…49 °C
MDL Number: MFCD10758067
CAS Number: 679839-39-5

Order Now

Specification Test Results

Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR