3-Nitro-2-picoline ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-3-nitropyridine

046603-5G
101876-238EA 36.92 USD
101876-238
3-Nitro-2-picoline ≥95%
3-Nitro-2-picoline

Hazard: Irritant

Formula: C₆H₆N₂O₂
MW: 138.1240 g/mol
Melting Pt: 32…33 °C
MDL Number: MFCD04114137
CAS Number: 18699-87-1

Order Now

Specification Test Results

Melting point 32-33°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR