7-Bromo-4-methyl-5-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

046596-500MG
101876-216EA 688.85 USD
101876-216
7-Bromo-4-methyl-5-nitro-1H-indazole ≥95%
7-Bromo-4-methyl-5-nitro-1H-indazole

Hazard: Irritant

Formula: C₈H₆BrN₃O₂
Melting Pt: 204…206 °C
MDL Number: MFCD09607937
CAS Number: 952183-46-9

Order Now

Specification Test Results

Melting point 204-206°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR