7-Chloro-1H-indazole ≥95%
Supplier: Matrix Scientific
Synonyms:
7-Chloroindazole
046591-1G
101876-192EA
184.62
USD
101876-192
7-Chloro-1H-indazole ≥95%
7-Chloro-1H-indazole
Hazard: Irritant
Formula:
C₇H₅ClN₂ MW: 152.5810 g/mol Melting Pt: 135…137 °C |
MDL Number:
MFCD07368214 CAS Number: 37435-12-4 |
Specification Test Results
Melting point | 135-137°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...