Methyl thieno[3,2-b]thiophene-2-carboxylate ≥95%
Supplier: Matrix Scientific
046590-500MG
046590-5G
046590-1G
101876-186EA
467.69
USD
101876-186
101876-184
101876-188
Methyl thieno[3,2-b]thiophene-2-carboxylate ≥95%
Methyl thieno[3,2-b]thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₈H₆O₂S₂ Melting Pt: 93…95 °C |
MDL Number:
MFCD01934498 CAS Number: 98800-10-3 |
Specification Test Results
Melting point | 93-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...