2-(1H-Indazol-6-yl)tetrahydro-1H-1λ~6~-isothiazole-1,1-dione ≥95%

Supplier: Matrix Scientific

Synonyms: 6-(1,1-Dioxoisothiazolidin-2-yl)-1H-indazole

046589-500MG 046589-1G
101876-182EA 698.46 USD
101876-182 101876-180
2-(1H-Indazol-6-yl)tetrahydro-1H-1λ~6~-isothiazole-1,1-dione ≥95%
2-(1H-Indazol-6-yl)tetrahydro-1H-1lambda~6~-isothiazole-1,1-dione

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₂S
MW: 237.28 g/mol
Melting Pt: 201…202 °C
MDL Number: MFCD09607932
CAS Number: 952183-42-5

Order Now

Specification Test Results

Melting point 201-202°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR