2-Phenyl-1H-imidazole-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
046568-1G
101876-080EA
392.31
USD
101876-080
2-Phenyl-1H-imidazole-4-carbaldehyde ≥95%
2-Phenyl-1H-imidazole-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₀H₈N₂O MW: 172.19 g/mol Melting Pt: 168…170 °C |
MDL Number:
MFCD00173729 CAS Number: 68282-47-3 |
Specification Test Results
Melting point | 168-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...