1-(4-Chlorobenzyl)-1H-1,2,3-triazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

046567-500MG 046567-1G
101876-076EA 522.1 USD
101876-076 101876-074
1-(4-Chlorobenzyl)-1H-1,2,3-triazole-4-carbaldehyde ≥95%
1-(4-Chlorobenzyl)-1H-1,2,3-triazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₈ClN₃O
MW: 221.65 g/mol
Melting Pt: 92…94 °C
MDL Number: MFCD09607915
CAS Number: 952183-30-1

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR