[1-(4-Methylbenzyl)-1H-1,2,3-triazol-4-yl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Hydroxymethyl)-1-(4-methylbenzyl)-1H-1,2,3-triazole

046562-1G 046562-500MG
101876-054EA 280.6 USD
101876-054 101876-056
[1-(4-Methylbenzyl)-1H-1,2,3-triazol-4-yl]methanol ≥95%
[1-(4-Methylbenzyl)-1H-1,2,3-triazol-4-yl]methanol

Hazard: Irritant

Formula: C₁₁H₁₃N₃O
MW: 203.25 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD09607911
CAS Number: 952183-26-5

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR