3-(4-Bromo-2-fluorophenyl)-3-oxopropanenitrile ≥95%
Supplier: Matrix Scientific
046557-1G
046557-500MG
046557-5G
101876-032EA
206.15
USD
101876-032
101876-036
101876-034
3-(4-Bromo-2-fluorophenyl)-3-oxopropanenitrile ≥95%
3-(4-Bromo-2-fluorophenyl)-3-oxopropanenitrile
Hazard: Irritant
Formula:
C₉H₅BrFNO Melting Pt: 116…118 °C |
MDL Number:
MFCD09607907 CAS Number: 267880-92-2 |
Specification Test Results
Melting point | 116-118°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...