2-Amino-5-bromonicotinaldehyde ≥95%

Supplier: Matrix Scientific

046553-500MG 046553-1G
101876-018EA 253 USD
101876-018 101876-016
2-Amino-5-bromonicotinaldehyde ≥95%
2-Amino-5-bromonicotinaldehyde

Hazard: Irritant

Formula: C₆H₅BrN₂O
Melting Pt: 167…169 °C
MDL Number: MFCD09607904
CAS Number: 206997-15-1

Order Now

Specification Test Results

Melting point 167-169°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR