Naphtho[2,1-b]furan-2(1H)-one ≥95%
Supplier: Matrix Scientific
046522-1G
046522-5G
046522-500MG
101875-878EA
806.15
USD
101875-878
101875-874
101875-876
Naphtho[2,1-b]furan-2(1H)-one ≥95%
Naphtho[2,1-b]furan-2(1H)-one
Hazard: Irritant
Formula:
C₁₂H₈O₂ Melting Pt: 104…106 °C |
MDL Number:
MFCD08443985 CAS Number: 4352-63-0 |
Specification Test Results
Melting point | 104-106°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...