2-Methyl-7-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
046520-5G
046520-1G
046520-500MG
101875-864EA
467.69
USD
101875-864
101875-866
101875-862
2-Methyl-7-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid ≥95%
2-Methyl-7-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₈N₂O₆ MW: 252.19 g/mol Melting Pt: 173 °C |
MDL Number:
MFCD08443984 CAS Number: 154365-43-2 |
Specification Test Results
Melting point | 173°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...