2-Methyl-6-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

046519-500MG 046519-5G 046519-1G
101875-856EA 280.6 USD
101875-856 101875-858 101875-860
2-Methyl-6-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid ≥95%
2-Methyl-6-nitro-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₆
MW: 252.19 g/mol
Melting Pt: 176 °C
MDL Number: MFCD08443983
CAS Number: 154365-44-3

Order Now

Specification Test Results

Melting point 176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR