4-Bromo-2-chloro-6-[(dimethylamino)methyl]benzenol ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Bromo-2-chloro-6-[(dimethylamino)methyl]phenol
046516-1G
046516-5G
046516-500MG
101875-846EA
232.31
USD
101875-846
101875-848
101875-844
4-Bromo-2-chloro-6-[(dimethylamino)methyl]benzenol ≥95%
4-Bromo-2-chloro-6-[(dimethylamino)methyl]benzenol
Hazard: Irritant
Formula:
C₉H₁₁BrClNO MW: 264.56 g/mol Melting Pt: 69…71 °C |
MDL Number:
MFCD08443982 CAS Number: 924868-93-9 |
Specification Test Results
Melting point | 69-71°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...