1-(4-Isopropylphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

046514-500MG 046514-1G
101875-834EA 747.69 USD
101875-834 101875-836
1-(4-Isopropylphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
1-(4-Isopropylphenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₂
Melting Pt: 174…177 °C
MDL Number: MFCD11501767
CAS Number: 1171764-30-9

Order Now

Specification Test Results

Melting point 174-177°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR