4'-Nitro[1,1'-biphenyl]-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

046493-5G 046493-1G 046493-500MG
101875-724EA 262.2 USD
101875-724 101875-726 101875-722
4'-Nitro[1,1'-biphenyl]-2-carbaldehyde ≥95%
4'-Nitro[1,1'-biphenyl]-2-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₉NO₃
MW: 227.22 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD08443965
CAS Number: 924868-84-8

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR