(4-Pyrazol-1-ylphenyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Pyrazol-1-yl-phenyl)methanol

046492-1G 046492-5G
101875-718EA 135.38 USD
101875-718 101875-720
(4-Pyrazol-1-ylphenyl)methanol ≥95%
(4-Pyrazol-1-ylphenyl)methanol

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MW: 174.2000 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD02682057
CAS Number: 143426-49-7

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR