3',4'-Dichloro[1,1'-biphenyl]-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
046490-5G
046490-500MG
046490-1G
101875-706EA
121.54
USD
101875-706
101875-708
101875-710
3',4'-Dichloro[1,1'-biphenyl]-2-carbaldehyde ≥95%
3',4'-Dichloro[1,1'-biphenyl]-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₃H₈Cl₂O MW: 251.11 g/mol Melting Pt: 95…97 °C |
MDL Number:
MFCD08443964 CAS Number: 924868-83-7 |
Specification Test Results
Melting point | 95-97°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...