Methyl-4-(1-hydroxypropyl)-1-methyl-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

046481-500MG
101875-664EA 747.69 USD
101875-664
Methyl-4-(1-hydroxypropyl)-1-methyl-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-(1-hydroxypropyl)-1-methyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₅NO₃
MW: 197.24 g/mol
Melting Pt: 142…144 °C
MDL Number: MFCD12025864
CAS Number: 1135283-27-0

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR