tert-Butyl-2-amino-7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-amino-7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate

046459-500MG 046459-1G
101875-580EA 556.92 USD
101875-580 101875-582
tert-Butyl-2-amino-7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate ≥95%
tert-Butyl-2-amino-7,8-dihydropyrido[4,3-d]pyrimidine-6(5H)-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₈N₄O₂
MW: 250.3000 g/mol
Melting Pt: 170…172 °C
MDL Number: MFCD08447402
CAS Number: 869198-95-8

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR