Methyl-4-formyl-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-formyl-1H-pyrrole-2-carboxylate

046446-500MG 046446-1G
101875-530EA 921.54 USD
101875-530 101875-532
Methyl-4-formyl-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-formyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 124…125 °C
MDL Number: MFCD02179575
CAS Number: 40611-79-8

Order Now

Specification Test Results

Melting point 124-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR