Methyl-2-(3,5-dibromo-4-nitro-1H-pyrazol-1-yl)acetate ≥95%
Supplier: Matrix Scientific
046441-1G
046441-500MG
101875-512EA
698.46
USD
101875-512
101875-510
Methyl-2-(3,5-dibromo-4-nitro-1H-pyrazol-1-yl)acetate ≥95%
Methyl-2-(3,5-dibromo-4-nitro-1H-pyrazol-1-yl)acetate
Hazard: Irritant
Formula:
C₆H₅Br₂N₃O₄ Melting Pt: 135…136 °C |
MDL Number:
MFCD09972230 CAS Number: 1172351-19-7 |
Specification Test Results
Melting point | 135-136°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...