2-[5-Cyano-6-methyl-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (5-Cyano-3,4-dihydro-2,4-dioxo-6-methylpyrimidin-1(2H)-yl)acetic acid

046423-1G 046423-500MG
101875-430EA 280.6 USD
101875-430 101875-432
2-[5-Cyano-6-methyl-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid ≥95%
2-[5-Cyano-6-methyl-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]acetic acid

Hazard: Irritant

Formula: C₈H₇N₃O₄
MW: 209.16 g/mol
Melting Pt: 271…274 °C
MDL Number: MFCD09607900
CAS Number: 5900-45-8

Order Now

Specification Test Results

Melting point 271-274°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR