Ethyl-6-iodo-8-methylimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Supplier: Matrix Scientific
046420-1G
046420-500MG
101875-418EA
556.92
USD
101875-418
101875-420
Ethyl-6-iodo-8-methylimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 6-iodo-8-methylimidazo[1,2-a]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₁IN₂O₂ Melting Pt: 134…136 °C |
MDL Number:
MFCD09152737 CAS Number: 952183-17-4 |
Specification Test Results
Melting point | 134-136°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...