4-Fluoro-2-methyl-6-nitroaniline ≥95%

Supplier: Matrix Scientific

046409-1G
101875-378EA 11.5 USD
101875-378
4-Fluoro-2-methyl-6-nitroaniline ≥95%
4-Fluoro-2-methyl-6-nitroaniline

Hazard: Irritant

Formula: C₇H₇FN₂O₂
Melting Pt: 98…100 °C
MDL Number: MFCD09152778
CAS Number: 147285-87-8

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR