2-(4-Fluorophenyl)-2-oxoethyl acetate ≥95%
Supplier: Matrix Scientific
046392-500MG
046392-1G
046392-5G
101875-306EA
532.31
USD
101875-306
101875-302
101875-304
2-(4-Fluorophenyl)-2-oxoethyl acetate ≥95%
2-(4-Fluorophenyl)-2-oxoethyl acetate
Hazard: Irritant
Formula:
C₁₀H₉FO₃ Melting Pt: 52…54 °C |
MDL Number:
MFCD09152727 CAS Number: 366-78-9 |
Specification Test Results
Melting point | 52-54°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...