2-(4-Bromo-2-chlorophenoxy)acetonitrile ≥95%
Supplier: Matrix Scientific
046386-5G
046386-500MG
046386-1G
101875-272EA
532.31
USD
101875-272
101875-270
101875-268
2-(4-Bromo-2-chlorophenoxy)acetonitrile ≥95%
2-(4-Bromo-2-chlorophenoxy)acetonitrile
Hazard: Irritant
Formula:
C₈H₅BrClNO Melting Pt: 49…51 °C |
MDL Number:
MFCD09152774 CAS Number: 951918-41-5 |
Specification Test Results
Melting point | 49-51°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...