2-(2-Chloro-4-fluorophenoxy)ethanethioamide ≥95%

Supplier: Matrix Scientific

046385-500MG 046385-1G
101875-264EA 556.92 USD
101875-264 101875-266
2-(2-Chloro-4-fluorophenoxy)ethanethioamide ≥95%
2-(2-Chloro-4-fluorophenoxy)ethanethioamide

Hazard: Irritant

Formula: C₈H₇ClFNOS
Melting Pt: 99…101 °C
MDL Number: MFCD09152726
CAS Number: 952183-08-3

Order Now

Specification Test Results

Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR