2-[4-(Trifluoromethoxy)phenoxy]ethanethioamide ≥95%

Supplier: Matrix Scientific

046380-1G 046380-500MG
101875-250EA 698.46 USD
101875-250 101875-248
2-[4-(Trifluoromethoxy)phenoxy]ethanethioamide ≥95%
2-[4-(Trifluoromethoxy)phenoxy]ethanethioamide

Hazard: Irritant

Formula: C₉H₈F₃NO₂S
Melting Pt: 97…99 °C
MDL Number: MFCD09152725
CAS Number: 952183-07-2

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR