3-(3,5-Dimethoxyphenyl)-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-3-(3,5-dimethoxyphenyl)-1H-pyrazole

046353-500MG 046353-1G 046353-5G
101875-120EA 241.5 USD
101875-120 101875-118 101875-122
3-(3,5-Dimethoxyphenyl)-1H-pyrazol-5-amine ≥95%
3-(3,5-Dimethoxyphenyl)-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₂
Melting Pt: 145…147 °C
MDL Number: MFCD08443954
CAS Number: 569655-99-8

Order Now

Specification Test Results

Melting point 145-147°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR