3-(3-Methoxyphenyl)-3-oxopropanenitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Methoxybenzoylacetonitrile
046352-500MG
046352-5G
046352-1G
101875-116EA
192.31
USD
101875-116
101875-112
101875-114
3-(3-Methoxyphenyl)-3-oxopropanenitrile ≥95%
3-(3-Methoxyphenyl)-3-oxopropanenitrile
Hazard: Irritant
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Melting Pt: 89…91 °C |
MDL Number:
MFCD01566517 CAS Number: 21667-60-7 |
Specification Test Results
Melting point | 89-91°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...