5-Chloro-6-(dimethylamino)-2-methyl-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

046275-500MG
101874-914EA 615.38 USD
101874-914
5-Chloro-6-(dimethylamino)-2-methyl-4-pyrimidinol ≥95%
5-Chloro-6-(dimethylamino)-2-methyl-4-pyrimidinol

Hazard: Irritant

Formula: C₇H₁₀ClN₃O
Melting Pt: 231…233 °C
MDL Number: MFCD11840986
CAS Number: 1135283-12-3

Order Now

Specification Test Results

Melting point 231-233°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR