6-(Trifluoromethyl)-1-benzothiophene-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
046264-1G
046264-500MG
101874-876EA
920
USD
101874-876
101874-874
6-(Trifluoromethyl)-1-benzothiophene-2-carbaldehyde ≥95%
6-(Trifluoromethyl)-1-benzothiophene-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₀H₅F₃OS MW: 230.21 g/mol Melting Pt: 54…56 °C |
MDL Number:
MFCD11840979 CAS Number: 863118-49-4 |
Specification Test Results
Melting point | 54-56°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...