3-Methoxy-4-(trifluoromethyl)phenylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methoxy-4-(trifluoromethyl)aniline

046260-500MG
101874-860EA 178.25 USD
101874-860
3-Methoxy-4-(trifluoromethyl)phenylamine ≥95%
3-Methoxy-4-(trifluoromethyl)phenylamine

Hazard: Irritant

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Melting Pt: 49…50 °C
MDL Number: MFCD11840978
CAS Number: 106877-20-7

Order Now

Specification Test Results

Melting point 49-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR