tert-Butyl-2-(2-chloroacetyl)-1,2-diazepane-1-carboxylate ≥95%

Supplier: Matrix Scientific

046258-500MG
101874-856EA 558.9 USD
101874-856
tert-Butyl-2-(2-chloroacetyl)-1,2-diazepane-1-carboxylate ≥95%
tert-Butyl-2-(2-chloroacetyl)-1,2-diazepane-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₁ClN₂O₃
MW: 276.77 g/mol
Melting Pt: 72…73 °C
MDL Number: MFCD11840976
CAS Number: 1135283-02-1

Order Now

Specification Test Results

Melting point 72-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR