2-Methyl-6-(methylamino)-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

046249-500MG
101874-824EA 375.38 USD
101874-824
2-Methyl-6-(methylamino)-4-pyrimidinol ≥95%
2-Methyl-6-(methylamino)-4-pyrimidinol

Hazard: Irritant

Formula: C₆H₉N₃O
MW: 139.16 g/mol
Melting Pt: 288…291 °C
MDL Number: MFCD11840972
CAS Number: 857414-73-4

Order Now

Specification Test Results

Melting point 288-291°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR