4-(4-Methylpiperazin-1-yl)benzoic acid hydrochloride ≥95%

Supplier: Matrix Scientific

046227-500MG
101874-738EA 278.46 USD
101874-738
4-(4-Methylpiperazin-1-yl)benzoic acid hydrochloride ≥95%
4-(4-Methylpiperazin-1-yl)benzoic acid hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂
Melting Pt: 245 °C (dec.)
MDL Number: MFCD06641474
CAS Number: 289044-60-6

Order Now

Specification Test Results

Melting point 245°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR