tert-Butyl-4-(6-chloro-5-formyl-4-pyrimidinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

046211-1G 046211-5G 046211-500MG
101874-690EA 802.7 USD
101874-690 101874-688 101874-686
tert-Butyl-4-(6-chloro-5-formyl-4-pyrimidinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(6-chloro-5-formyl-4-pyrimidinyl)tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₉ClN₄O₃
Melting Pt: 120…122 °C
MDL Number: MFCD09972225
CAS Number: 1017782-55-6

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR