2-[2-(Methylsulfonyl)phenoxy]acetic acid ≥95%

Supplier: Matrix Scientific

046198-1G
101874-644EA 460 USD
101874-644
2-[2-(Methylsulfonyl)phenoxy]acetic acid ≥95%
2-[2-(Methylsulfonyl)phenoxy]acetic acid

Hazard: Irritant

Formula: C₉H₁₀O₅S
Melting Pt: 181…182 °C
MDL Number: MFCD09972219
CAS Number: 1017782-54-5

Order Now

Specification Test Results

Melting point 181-182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR