(5-Chloro-1,3-benzoxazol-2-yl)-N-methylmethanamine ≥95%

Supplier: Matrix Scientific

046193-500MG
101874-620EA 747.69 USD
101874-620
(5-Chloro-1,3-benzoxazol-2-yl)-N-methylmethanamine ≥95%
(5-Chloro-1,3-benzoxazol-2-yl)-N-methylmethanamine

Hazard: Irritant

Formula: C₉H₉ClN₂O
MW: 196.64 g/mol
Melting Pt: 139…141 °C
MDL Number: MFCD09972215
CAS Number: 1017782-51-2

Order Now

Specification Test Results

Melting point 139-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR