Ethyl-1-(4-amino-3-methoxyphenyl)-4-piperidinecarboxylate ≥95%
Supplier: Matrix Scientific
046186-1G
046186-500MG
101874-584EA
280.6
USD
101874-584
101874-586
Ethyl-1-(4-amino-3-methoxyphenyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(4-amino-3-methoxyphenyl)-4-piperidinecarboxylate
Hazard: Irritant
Formula:
C₁₅H₂₂N₂O₃ Melting Pt: 76…77 °C |
MDL Number:
MFCD09972211 CAS Number: 1017782-48-7 |
Specification Test Results
Melting point | 76-77°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...