6-Methyl-5-(1-pyrrolidinylmethyl)-2,4(1H,3H)-pyrimidinedione ≥95%
Supplier: Matrix Scientific
046177-1G
046177-5G
046177-500MG
101874-540EA
532.31
USD
101874-540
101874-538
101874-536
6-Methyl-5-(1-pyrrolidinylmethyl)-2,4(1H,3H)-pyrimidinedione ≥95%
6-Methyl-5-(1-pyrrolidinylmethyl)-2,4(1H,3H)-pyrimidinedione
Hazard: Irritant
Formula:
C₁₀H₁₅N₃O₂ Melting Pt: >300 °C |
MDL Number:
MFCD10758054 CAS Number: 1033463-16-9 |
Specification Test Results
Melting point |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...