N'-(1H-Indazol-5-yl)-N,N-dimethyliminoformamide ≥95%

Supplier: Matrix Scientific

Synonyms: (E)-N'-(1H-Indazol-5-yl)-N,N-dimethylmethanimidamide

046174-500MG 046174-5G 046174-1G
101874-526EA 602.6 USD
101874-526 101874-524 101874-522
N'-(1H-Indazol-5-yl)-N,N-dimethyliminoformamide ≥95%
N'-(1H-Indazol-5-yl)-N,N-dimethyliminoformamide

Hazard: Irritant

Formula: C₁₀H₁₂N₄
MW: 188.23 g/mol
Melting Pt: 129…130 °C
MDL Number: MFCD09152720
CAS Number: 952183-04-9

Order Now

Specification Test Results

Melting point 129-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR