Ethyl-4-{2-[4-(methylsulfonyl)phenyl]-2-oxoethyl}tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 4-{2-[4-(methylsulphonyl)phenyl]-2-oxoethyl}piperazine-1-carboxylate
046156-5G
046156-500MG
046156-1G
101874-440EA
532.31
USD
101874-440
101874-438
101874-436
Ethyl-4-{2-[4-(methylsulfonyl)phenyl]-2-oxoethyl}tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Ethyl-4-{2-[4-(methylsulfonyl)phenyl]-2-oxoethyl}tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₂₂N₂O₅S MW: 354.43 g/mol Melting Pt: 152…154 °C |
MDL Number:
MFCD08689706 CAS Number: 924869-22-7 |
Specification Test Results
Melting point | 152-154°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...