5-(4-Bromophenoxy)-2-furoic acid ≥95%
Supplier: Matrix Scientific
046143-5G
046143-1G
046143-500MG
101874-364EA
300
USD
101874-364
101874-368
101874-366
5-(4-Bromophenoxy)-2-furoic acid ≥95%
5-(4-Bromophenoxy)-2-furoic acid
Hazard: Irritant
Formula:
C₁₁H₇BrO₄ Melting Pt: 173…175 °C |
MDL Number:
MFCD09607931 CAS Number: 73420-68-5 |
Specification Test Results
Melting point | 173-175°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...