1-Methyl-4-nitro-1H-indazole ≥95%
Supplier: Matrix Scientific
046139-500MG
101874-348EA
1109.23
USD
101874-348
1-Methyl-4-nitro-1H-indazole ≥95%
1-Methyl-4-nitro-1H-indazole
Hazard: Irritant
Formula:
C₈H₇N₃O₂ MW: 177.1600 g/mol Melting Pt: 141…142 °C |
MDL Number:
MFCD09607929 CAS Number: 26120-43-4 |
Specification Test Results
Melting point | 141-142°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...