1-Methyl-1H-indazol-6-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Amino-1-methyl-1H-indazole

046137-1G
101874-340EA 153.85 USD
101874-340
1-Methyl-1H-indazol-6-amine ≥95%
1-Methyl-1H-indazol-6-amine

Hazard: Irritant

Formula: C₈H₉N₃
MW: 147.18 g/mol
Melting Pt: 172…173 °C
MDL Number: MFCD00456076
CAS Number: 74728-65-7

Order Now

Specification Test Results

Melting point 172-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR