1-(4-Amino-3-methoxyphenyl)-1-ethanone ≥97%
Supplier: Matrix Scientific
Synonyms:
4'-Amino-3'-methoxyacetophenone
046132-1G
046132-500MG
046132-5G
101874-308EA
166.15
USD
101874-308
101874-312
101874-310
1-(4-Amino-3-methoxyphenyl)-1-ethanone ≥97%
1-(4-Amino-3-methoxyphenyl)-1-ethanone
Hazard: Irritant
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol Melting Pt: 90…92 °C |
MDL Number:
MFCD08443961 CAS Number: 22106-40-7 |
Specification Test Results
Melting point | 90-92°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...