4-Isopropylbenzenecarbothioamide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Propan-2-yl)benzene-1-carbothioamide

046122-500MG 046122-1G
101874-264EA 262.2 USD
101874-264 101874-262
4-Isopropylbenzenecarbothioamide ≥95%
4-Isopropylbenzenecarbothioamide

Hazard: Irritant

Formula: C₁₀H₁₃NS
MW: 179.29 g/mol
Melting Pt: 150…152 °C
MDL Number: MFCD05264094
CAS Number: 53515-20-1

Order Now

Specification Test Results

Melting point 150-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR